Anti-GPR20 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB16197
10684-550EA 653.85 USD
10684-550
Anti-GPR20 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of GPR20.

Recommended Dilutions: Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 20 ?g/ml

Type: Primary
Antigen: GPR20
Clonality: Unconjugated
Clone: Polyclonal
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Bovine, Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR