Anti-PRDX1 Mouse Monoclonal Antibody [clone: AT1D8]

Supplier: Abnova
MAB7559
10730-048EA 319 USD
10730-048
Anti-PRDX1 Mouse Monoclonal Antibody [clone: AT1D8]
Mouse monoclonal antibody raised against recombinant PRDX1.

Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: PRDX1
Clonality: Monoclonal
Clone: AT1D8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR