Anti-SNX18 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB23168
10738-886EA 527.69 USD
10738-886
Anti-SNX18 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against recombinant SNX18.

Recommended Dilutions: Immunohistochemistry: 1:20-1:50; Western Blot: 1:250-1:500

Type: Primary
Antigen: SNX18
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR