Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Salmonella Mouse Monoclonal Antibody [clone: 5D12A]

Supplier: Abnova
MAB7718
10677-222
Anti-Salmonella Mouse Monoclonal Antibody [clone: 5D12A]
Mouse monoclonal antibody raised against Salmonella.

Type: Primary
Antigen: Salmonella
Clonality: Unconjugated
Clone: Monoclonal
Conjugation: 5D12A
Epitope:
Host: IgG1
Isotype: Mouse
Reactivity: Bacteria

Order Now


 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty