Anti-ADAMTS4 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB16220
10683-150EA 653.85 USD
10683-150
Anti-ADAMTS4 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of ADAMTS4.

Recommended Dilutions: Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 20 ?g/ml

Type: Primary
Antigen: ADAMTS4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Dog, Horse, Human, Orangutan
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR