Anti-Oval Cell Marker Rat Monoclonal Antibody [clone: OC2-3C5]

Supplier: Abnova
MAB7742
10728-000EA 653.85 USD
10728-000
Anti-Oval Cell Marker Rat Monoclonal Antibody [clone: OC2-3C5]
Antibodies
Rat monoclonal antibody raised against native oval cell marker.

Recommended Dilutions: Flow Cytometry: 1:50-1:100; Immunohistochemistry (Frozen sections): 1:100

Type: Primary
Antigen: Oval Cell Marker
Clonality: Unconjugated
Clone: Monoclonal
Conjugation: OC2-3C5
Epitope:
Host: IgG1
Isotype: Rat
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR