Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PLEKHM2 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB16862
10729-440
Anti-PLEKHM2 Rabbit Polyclonal Antibody
Rabbit polyclonal antibody raised against synthetic peptide of PLEKHM2.

Recommended Dilutions: Western Blot: 0.5-1 μg/ml

Type: Primary
Antigen: PLEKHM2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat

Order Now


 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty