Anti-Oval Cell Marker Rat Monoclonal Antibody [clone: OC2-2F3]

Supplier: Abnova
MAB7755
10733-278EA 653.85 USD
10733-278
Anti-Oval Cell Marker Rat Monoclonal Antibody [clone: OC2-2F3]
Antibodies
Rat monoclonal antibody raised against native oval cell marker.

Recommended Dilutions: Flow Cytometry: 1:50-1:100; Immunohistochemistry (Frozen sections): 1:100

Type: Primary
Antigen: Oval Cell Marker
Clonality: Unconjugated
Clone: Monoclonal
Conjugation: OC2-2F3
Epitope:
Host: IgG1
Isotype: Rat
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR