Anti-Influenza A virus Mouse Monoclonal Antibody [clone: 4A1]

Supplier: Abnova
MAB8540
10628-214EA 515 USD
10628-214
Anti-Influenza A virus Mouse Monoclonal Antibody [clone: 4A1]
Antibodies
Mouse monoclonal antibodt raised against Influenza A virus.

Type: Primary
Antigen: Influenza A virus
Clonality: Unconjugated
Clone: Monoclonal
Conjugation: 4A1
Epitope:
Host: IgG2a
Isotype: Mouse
Reactivity: Viruses
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR