Anti-CABS1 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB16867
10704-348EA 527.69 USD
10704-348
Anti-CABS1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of C4orf35.

Recommended Dilutions: Western Blot: 1-2 ?g/ml

Type: Primary
Antigen: CABS1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR