Anti-IL4 Rat Monoclonal Antibody

MAB8546
10719-260EA 421.54 USD
10719-260
Anti-IL4 Rat Monoclonal Antibody
Rat monoclonal antibody raised against Il4.

Recommended Dilutions: ELISA: 1:1000-1:2000; Western Blot: 2-5 ?g/ml

Type: Primary
Antigen: IL4
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR