Anti-CCDC47 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB16870
10705-554EA 527.69 USD
10705-554
Anti-CCDC47 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of CCDC47.

Recommended Dilutions: Western Blot: 1-2 ?g/ml

Type: Primary
Antigen: CCDC47
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR