Anti-IL25 Mouse Monoclonal Antibody [clone: SB129a]

Supplier: Abnova
MAB8555
10735-238EA 527.69 USD
10735-238
Anti-IL25 Mouse Monoclonal Antibody [clone: SB129a]
Antibodies
Mouse monoclonal antibody raised against full length recombinant IL25.

Recommended Dilutions: ELISA: 0.1-1 ?g/ml; Immunostaining (2-5 ?g/ml

Type: Primary
Antigen: IL25
Clonality: Monoclonal
Clone: SB129a
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR