Anti-KCNIP1 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB16878
10721-938EA 426.15 USD
10721-938
Anti-KCNIP1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of Kcnip1.

Recommended Dilutions: Enzyme Immunoassay (1:300000; Western Blot: 1:4000

Type: Primary
Antigen: KCNIP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR