Anti-JAK2 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB16879
10719-966EA 426.15 USD
10719-966
Anti-JAK2 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of JAK2.

Recommended Dilutions: Western Blot: 1:2000; ELISA: 1:64000

Type: Primary
Antigen: JAK2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR