Anti-OLAH Rabbit Polyclonal Antibody

Supplier: Abnova
PAB23498
10727-938EA 527.69 USD
10727-938
Anti-OLAH Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against recombinant OLAH.

Recommended Dilutions: Immunohistochemistry: 1:10-1:20; Western Blot: 1:250-1:500

Type: Primary
Antigen: OLAH
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR