Anti-FIH Mouse Monoclonal Antibody [clone: FIH 162c]

Supplier: Abnova
MAB8563
10714-774EA 415 USD
10714-774
Anti-FIH Mouse Monoclonal Antibody [clone: FIH 162c]
Antibodies
Mouse monoclonal antibody raised against full length recombinant FIH.

Recommended Dilutions: Western Blot: 10 ?g/ml

Type: Primary
Antigen: FIH
Clonality: Unconjugated
Clone: Monoclonal
Conjugation: FIH 162c
Epitope:
Host:
Isotype: Mouse
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR