Anti-BCL2 0 Polyclonal Antibody

Supplier: Abnova
PAB16882
10674-310EA 527.69 USD
10674-310
Anti-BCL2 0 Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic phosphopeptide of BCL2.

Recommended Dilutions: ELISA: 1:2000-1:100000

Type: Primary
Antigen: BCL2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: 0
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR