Anti-GABRB3 Mouse Monoclonal Antibody [clone: S87-25]

Supplier: Abnova
MAB8567
10720-442EA 653.85 USD
10720-442
Anti-GABRB3 Mouse Monoclonal Antibody [clone: S87-25]
Antibodies
Mouse monoclonal antibody raised against partial native Gabrb3.

Recommended Dilutions: Western Blot: 1 ?g/ml; Immunohistochemistry: 0.1-1.0 ?g/ml

Type: Primary
Antigen: Gabrb3
Clonality: Unconjugated
Clone: Monoclonal
Conjugation: S87-25
Epitope:
Host: IgG1
Isotype: Mouse
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR