Anti-IKBKG Polyclonal Antibody

Supplier: Abnova
PAB16885
10674-766EA 527.69 USD
10674-766
Anti-IKBKG Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic phosphopeptide of IKBKG .

Recommended Dilutions: ELISA: 1:100000-1:400000

Type: Primary
Antigen: IKBKG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR