Anti-RELA Polyclonal Antibody

Supplier: Abnova
PAB16887
10675-272EA 527.69 USD
10675-272
Anti-RELA Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic phosphopeptide of RELA.

Recommended Dilutions: ELISA: 1:50000-1:120000

Type: Primary
Antigen: RELA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR