Anti-DDX23 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB23506
10710-356EA 527.69 USD
10710-356
Anti-DDX23 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against recombinant DDX23.

Recommended Dilutions: Immunohistochemistry: 1:1000-1:2500; Western Blot: 1:250-1:500; Immunofluorescence: 1-4 ?g/ml

Type: Primary
Antigen: DDX23
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR