Anti-TRPV3 Polyclonal Antibody

Supplier: Abnova
PAB16890
10742-998EA 426.15 USD
10742-998
Anti-TRPV3 Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of Trpv3.

Recommended Dilutions: EIA: 1:5000; Western Blot: 1:2000

Type: Primary
Antigen: TRPV3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR