Anti-KCNIP4 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB16892
10721-952EA 426.15 USD
10721-952
Anti-KCNIP4 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of Kcnip4.

Recommended Dilutions: EIA: 1:300000; Western Blot: 1:1000

Type: Primary
Antigen: KCNIP4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR