Anti-MOK Mouse Monoclonal Antibody [clone: 1C5]

MAB7936
10685-962EA 1683.08 USD
10685-962
Anti-MOK Mouse Monoclonal Antibody [clone: 1C5]
Mouse monoclonal antibody raised against recombinant RAGE.

Recommended Dilutions: ELISA: 0.01 ?g/ml; Flow Cytometry: 1 ?g/ml ICC (1 ?g/ml; Western Blot: 0.1 ?g/ml

Type: Primary
Antigen: MOK
Clonality: Monoclonal
Clone: 1C5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR