Anti-TYRO3 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB15505
10743-548EA 653.85 USD
10743-548
Anti-TYRO3 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of TYRO3.

Recommended Dilutions: Immunocytochemistry: 1:200; Immunofluorescence: 1:200; Western Blot: 2 ?g/ml

Type: Primary
Antigen: TYRO3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR