Anti-Hepatitis C virus NS5b protein Mouse Monoclonal Antibody [clone: NS5B-6]

Supplier: Abnova
MAB7950
10720-814EA 426.15 USD
10720-814
Anti-Hepatitis C virus NS5b protein Mouse Monoclonal Antibody [clone: NS5B-6]
Antibodies
Mouse monoclonal antibody raised against recombinant hepatitis C virus NS5b protein.

Type: Primary
Antigen: Hepatitis C virus NS5b protein
Clonality: Unconjugated
Clone: Monoclonal
Conjugation: NS5B-6
Epitope:
Host: IgG2a kappa
Isotype: Mouse
Reactivity: Viruses
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR