Anti-LDLR Rabbit Polyclonal Antibody

Supplier: Abnova
PAB15520
10723-452EA 653.85 USD
10723-452
Anti-LDLR Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of LDLR.

Recommended Dilutions: Western Blot: 0.5 ?g/ml

Type: Primary
Antigen: LDLR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Primates
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR