Anti-FER Rabbit Polyclonal Antibody

Supplier: Abnova
PAB15526
10720-238EA 653.85 USD
10720-238
Anti-FER Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of FER.

Recommended Dilutions: Western Blot: 0.5 ?g/ml

Type: Primary
Antigen: FER
Clonality: Unconjugated
Clone: Polyclonal
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Dog, Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR