Anti-CCT2 Mouse Monoclonal Antibody [clone: F39P7F11]

Supplier: Abnova
MAB7972
10683-520EA 920 USD
10683-520
Anti-CCT2 Mouse Monoclonal Antibody [clone: F39P7F11]
Antibodies
Mouse monoclonal antibody raised against synthetic peptide of CCT2.

Type: Primary
Antigen: CCT2
Clonality: Monoclonal
Clone: F39P7F11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Frog, Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR