Anti-ADARB2 Rabbit Polyclonal Antibody

PAB22715
10692-308EA 510.77 USD
10692-308
Anti-ADARB2 Rabbit Polyclonal Antibody
Rabbit polyclonal antibody raised against recombinant ADARB2.

Recommended Dilutions: Immunohistochemistry: 1:200-1:500; Immunofluorescence: 1-4 ?g/ml

Type: Primary
Antigen: ADARB2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR