Anti-UEA1 Polyclonal Antibody

PAB15537
10677-952EA 521.54 USD
10677-952
Anti-UEA1 Polyclonal Antibody
Rabbit polyclonal antibody raised against native Ulex Europaeus Lectin 1.

Recommended Dilutions: Immunofluorescence: 1:500; Immunohistochemistry (Frozen sections): 1:500; Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 1:500

Type: Primary
Antigen: UEA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype:
Reactivity: Human, Plant
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR