Anti-HLA-Class I Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: W6/32]

Supplier: Abnova
MAB5083
10720-964EA 426.15 USD
10720-964
Anti-HLA-Class I Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: W6/32]
Antibodies
Mouse monoclonal antibody raised against native HLA-Class I.

Recommended Dilutions: Flow Cytometry: 1:300

Type: Primary
Antigen: HLA-Class I
Clonality: FITC (Fluorescein Isothiocyanate)
Clone: Monoclonal
Conjugation: W6/32
Epitope:
Host: IgG2a
Isotype: Mouse
Reactivity: Bovine, Cat, Human, Primates
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR