Anti-HLA-Class I Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: W6/32]

Supplier: Abnova
MAB5084
10720-966EA 527.69 USD
10720-966
Anti-HLA-Class I Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: W6/32]
Antibodies
Mouse monoclonal antibody raised against native HLA-Class I.

Recommended Dilutions: Flow Cytometry: 1:50

Type: Primary
Antigen: HLA-Class I
Clonality: PE (Phycoerythrin)
Clone: Monoclonal
Conjugation: W6/32
Epitope:
Host: IgG2a
Isotype: Mouse
Reactivity: Bovine, Cat, Human, Primates
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR