Anti-SPHK1 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB15559
10739-284EA 415 USD
10739-284
Anti-SPHK1 Rabbit Polyclonal Antibody
Rabbit polyclonal antibody raised against SPHK1.

Recommended Dilutions: Immunoprecipitation: 5-15 ?g/mg lysate; Western Blot: 1:2000-1:10000

Type: Primary
Antigen: SPHK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR