Anti-ruvB Rabbit Polyclonal Antibody

Supplier: Abnova
PAB15570
10732-130EA 426.15 USD
10732-130
Anti-ruvB Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against full length recombinant Escherichia coli RuvB.

Recommended Dilutions: Western Blot: 1:3000

Type: Primary
Antigen: ruvB
Clonality: Unconjugated
Clone: Polyclonal
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: E. coli
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR