Anti-HLA-G Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 87G]

Supplier: Abnova
MAB5094
10717-576EA 653.85 USD
10717-576
Anti-HLA-G Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 87G]
Antibodies
Mouse monoclonal antibody raised against HLA-G.

Recommended Dilutions: Flow Cytometry: 1:10

Type: Primary
Antigen: HLA-G
Clonality: Monoclonal
Clone: 87G
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR