Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Escherichia coli UmuD Rabbit Polyclonal Antibody

Supplier: Abnova
PAB15572
10743-820
Anti-Escherichia coli UmuD Rabbit Polyclonal Antibody
Rabbit polyclonal antibody raised against recombinant Escherichia coli UmuD.

Recommended Dilutions: Western Blot: 1:3000

Type: Primary
Antigen: Escherichia coli UmuD
Clonality: Unconjugated
Clone: Polyclonal
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: E. coli

Order Now


 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty