Anti-TNFRSF8 Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: MEM-268]

Supplier: Abnova
MAB5099
10742-178EA 406.15 USD
10742-178
Anti-TNFRSF8 Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: MEM-268]
Antibodies
Mouse monoclonal antibody raised against TNFRSF8.

Recommended Dilutions: Flow Cytometry: 20 ?l in human blood cells 100 ?l in whole blood or 10^6 cells in a suspension)

Type: Primary
Antigen: TNFRSF8
Clonality: Monoclonal
Clone: MEM-268
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host:
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR