Anti-CRP Mouse Monoclonal Antibody (Biotin) [clone: C7]

Supplier: Abnova
MAB5101
10709-124EA 426.15 USD
10709-124
Anti-CRP Mouse Monoclonal Antibody (Biotin) [clone: C7]
Antibodies
Mouse monoclonal antibody raised against CRP.

Type: Primary
Antigen: CRP
Clonality: Monoclonal
Clone: C7
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR