Anti-CERK Rabbit Polyclonal Antibody

Supplier: Abnova
PAB16373
10683-614EA 653.85 USD
10683-614
Anti-CERK Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of CERK.

Type: Primary
Antigen: CERK
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR