Anti-HARBI1 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB23326
10716-920EA 527.69 USD
10716-920
Anti-HARBI1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against recombinant HARBI1.

Recommended Dilutions: Immunohistochemistry: 1:500-1:1000; Western Blot: 1:250-1:500; Immunofluorescence: 1-4 ?g/ml

Type: Primary
Antigen: HARBI1
Clonality: Unconjugated
Clone: Polyclonal
Conjugation:
Epitope:
Host: IgG
Isotype: Rabbit
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR