Anti-FZD7 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB16379
10692-940EA 653.85 USD
10692-940
Anti-FZD7 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of FZD7.

Recommended Dilutions: Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 15 ?g/ml

Type: Primary
Antigen: FZD7
Clonality: Unconjugated
Clone: Polyclonal
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Bovine, Dog, Gorilla, Hamster, Human, Monkey, Mouse, Rabbit, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR