Anti-TRPS1 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB23333
10743-080EA 527.69 USD
10743-080
Anti-TRPS1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against recombinant TRPT1.

Recommended Dilutions: Immunohistochemistry: 1:1000-1:2500; Western Blot: 1:250-1:500

Type: Primary
Antigen: TRPS1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR