Anti-CD69 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: FN50]

Supplier: Abnova
MAB5115
10706-752EA 406.15 USD
10706-752
Anti-CD69 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: FN50]
Antibodies
Mouse monoclonal antibody raised against native CD69.

Recommended Dilutions: Flow Cytometry: 20 ?l in human blood cells 100 ?l in whole blood or 10^6 cells in a suspension)

Type: Primary
Antigen: CD69
Clonality: Monoclonal
Clone: FN50
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR