Anti-ATG2A Rabbit Polyclonal Antibody

Supplier: Abnova
PAB23335
10690-370EA 527.69 USD
10690-370
Anti-ATG2A Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against recombinant ATG2A.

Recommended Dilutions: Immunohistochemistry: 1:500-1:1000

Type: Primary
Antigen: ATG2A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR