Anti-MCAM Mouse Monoclonal Antibody (Biotin) [clone: c264]

Supplier: Abnova
MAB5851
10735-436EA 653.85 USD
10735-436
Anti-MCAM Mouse Monoclonal Antibody (Biotin) [clone: c264]
Antibodies
Mouse monoclonal antibody raised against native MCAM.

Recommended Dilutions: Flow Cytometry: 1 ?g/10^6 cells

Type: Primary
Antigen: MCAM
Clonality: Monoclonal
Clone: c264
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Chicken
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR