Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-THYN1 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB23340
Rabbit polyclonal antibody raised against recombinant THYN1.

Recommended Dilutions: Immunohistochemistry: 1:200-1:500; Western Blot: 1:250-1:500; Immunofluorescence: 1-4 μg/ml

Type: Primary
Antigen: THYN1
Clonality: Unconjugated
Clone: Polyclonal
Conjugation:
Epitope:
Host: IgG
Isotype: Rabbit
Reactivity: Human




Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
100 µL PAB23340 10741-256 Each (100µl) Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen Symbol THYN1
Antigen Name Thymocyte Nuclear Protein 1
Reactivity Human
Conjugation Unconjugated
Clonality Polyclonal
Antibody Type Primary
Host Rabbit
Isotype IgG
Western Blot Yes
ImmunoChemistry Yes
ImmunoFluorescence Yes
Antigen Synonyms HSPC144, MDS012, THY28KD, MY105, THY28
Gene ID 29087
Format Liquid
Cross Adsorption No
Immunogen Recombinant protein corresponding to amino acids of human THYN1.
Purification Antigen affinity purification
Sequence AFFYHSNCKEPGIAGLMKIVKEAYPDHTQFEKNNPHYDPSSKEDNPKWSMVDVQFVRMMKRFIPLAELKSYHQAHKATGGPL
Storage Buffer PBS, pH 7.5 (40% glycerol, 0.02% sodium azide)
Storage Temperature Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing.
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty