Anti-VSIG1 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: CT1]

Supplier: Abnova
MAB5856
10736-276EA 653.85 USD
10736-276
Anti-VSIG1 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: CT1]
Antibodies
Mouse monoclonal antibody raised against native VSIG1.

Recommended Dilutions: Flow Cytometry: 1 ?g/10^6 cells

Type: Primary
Antigen: VSIG1
Clonality: Monoclonal
Clone: CT1
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Chicken
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR