Anti-CRYL1 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB23654
10709-184EA 527.69 USD
10709-184
Anti-CRYL1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against recombinant CRYL1.

Recommended Dilutions: Immunohistochemistry: 1:1000-1:2500

Type: Primary
Antigen: CRYL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR